missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,4-Diazabicyclo[4.3.0]nonane, 98+%

Catalog No. p-7034608
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52295MD 250 mg
AAH5229503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52295MD Supplier Thermo Scientific Chemicals Supplier No. H52295MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93643-24-4 | C7H14N2 | 126.20 g/mol

(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 93643-24-4
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.20
MDL Number MFCD03787926
InChI Key FTTATHOUSOIFOQ-SSDOTTSWSA-N
Synonym s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane
PubChem CID 781249
SMILES C1C[C@@H]2CNCCN2C1

Specifications

Quantity 250 mg
UN Number UN3267
Solubility Information Slightly soluble in water.
IUPAC Name (8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
Formula Weight 126.2
Percent Purity ≥98%
Chemical Name or Material (S)-1,4-Diazabicyclo[4.3.0]nonane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only