missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Bromophenyl)ethylamine 98.0+%, TCI America™

Catalog No. B36745G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36745G 5 g
B367425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36745G Supplier TCI America Supplier No. B36745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27298-97-1
Molecular Formula C8H11BrN
Molecular Weight (g/mol) 201.09
MDL Number MFCD00066026
InChI Key SOZMSEPDYJGBEK-LURJTMIESA-O
Synonym s---1-4-bromophenyl ethylamine, s-1-4-bromophenyl ethanamine, s---4-bromo-alpha-phenylethylamine, s-1-4-bromophenyl ethylamine, 1s-1-4-bromophenyl ethanamine, s-1-4-bromophenyl-ethylamine, 1s-1-4-bromophenyl ethan-1-amine, s---1-4-bromo-phenyl-ethylamine, benzenemethanamine,4-bromo-a-methyl-, as, s-1-4-bromophenyl ethanamine-hcl
PubChem CID 852999
IUPAC Name (1S)-1-(4-bromophenyl)ethan-1-aminium
SMILES C[C@H]([NH3+])C1=CC=C(Br)C=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 200.08
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-1-(4-Bromophenyl)ethylamine