missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%

Catalog No. p-7046784
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1632403 1 g
AAL1632406 5 g
AAL1632414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1632403 Supplier Thermo Scientific Chemicals Supplier No. L1632403

CAS: 82796-69-8 | C9H13NO | 151.209 g/mol

Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.

Solubility
Soluble in water (10g/L).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Chemical Identifiers

CAS 82796-69-8
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671656
InChI Key CJWGCBRQAHCVHW-ZETCQYMHSA-N
Synonym s-1-3-methoxyphenyl ethanamine, s-1-3-methoxyphenyl ethylamine, 1s-1-3-methoxyphenyl ethanamine, s-3-1-aminoethyl anisole, 1s-1-3-methoxyphenyl ethan-1-amine, s---1-3-methoxyphenyl ethylamine, r-+-1-3-methoxyphenyl ethylamine, chipros, ee, ksc496s2j, s-3-methoxyphenyl ethylamine, 1s-1-3-methoxyphenyl ethylamine
PubChem CID 7020761
IUPAC Name (1S)-1-(3-methoxyphenyl)ethanamine
SMILES CC(C1=CC(=CC=C1)OC)N

Specifications

Melting Point 5°C
Density 1.023
Boiling Point 66°C (0.38 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.5335
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in water (10g/L).
Optical Rotation −27° (Neat)
Formula Weight 151.21
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-1-(3-Methoxyphenyl)ethylamine
Show More Show Less
Hazard Category H302-H314-H331-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 632-902-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only