missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2-Butanediol 98.0+%, TCI America™

Catalog No. B41191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41191G 1 g
B41195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41191G Supplier TCI America Supplier No. B41191G
Only null left

Chemical Identifiers

CAS 73522-17-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD09953764
InChI Key BMRWNKZVCUKKSR-BYPYZUCNSA-N
Synonym (S)-1,2-Butylene Glycol, (S)-1,2-Dihydroxybutane
PubChem CID 6993189
ChEBI CHEBI:52686
IUPAC Name (2S)-butane-1,2-diol
SMILES CCC(CO)O

Specifications

Color Yellow
Boiling Point 55°C
Quantity 1 g
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-1,2-Butanediol
EINECSNumber (2)-0235
TSCA No