CAS RN 73522-17-5
IUPAC Name:
butane-1,2-diol
Synonyms:
1,2-butanediol α-butylene glycol α-butyleneglycol 1,2-butylene glycol ethylethylene glycol 1,2-(dihydroxy)butane
Molecular Weight (g/mol):
90.12
Molecular Formula:
C4H10O2
InChi Key:
BMRWNKZVCUKKSR-UHFFFAOYNA-N
SMILES:
CCC(O)CO
1
–
1
of
1
results
1
(S)-1,2-Butanediol 98.0+%, TCI America™
CAS: 73522-17-5 Molecular Formula: C4H10O2 Molecular Weight (g/mol): 90.122 MDL Number: MFCD09953764 InChI Key: BMRWNKZVCUKKSR-BYPYZUCNSA-N Synonym: (S)-1,2-Butylene Glycol, (S)-1,2-Dihydroxybutane PubChem CID: 6993189 ChEBI: CHEBI:52686 IUPAC Name: (2S)-butane-1,2-diol SMILES: CCC(CO)O