missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 99%

Catalog No. p-7046981
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1702203 1 g
AAL1702206 5 g
AAL1702214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1702203 Supplier Thermo Scientific Chemicals Supplier No. L1702203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23357-52-0 | C10H13N | 147.221 g/mol

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 23357-52-0
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.221
MDL Number MFCD00671630
InChI Key JRZGPXSSNPTNMA-JTQLQIEISA-N
Synonym s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1,2,3,4-tetrahydro-1-naphthylamine, s-+-1,2,3,4-tetrahydro-1-naphthylamine, 1s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1-amino-1,2,3,4-tetrahydronaphthalene, s-+-1-aminotetraline, chembl39537, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1s, s-1,2,3,4-tetrahydro-naphthalen-1-ylamine, s-1-amino-1,2,3,4-tetrahydro-naphthalene
PubChem CID 7058074
IUPAC Name (1S)-1,2,3,4-tetrahydronaphthalen-1-amine
SMILES C1CC(C2=CC=CC=C2C1)N

Specifications

Density 1.035
Boiling Point 118°C to 120°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5645
Quantity 1 g
Beilstein 2360279
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 147.22
Percent Purity ≥99%
Assay ee 99%
Chemical Name or Material (S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only