missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 99%
Description
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 1.035 |
| Boiling Point | 118°C to 120°C (8 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.5645 |
| Quantity | 5 g |
| Beilstein | 2360279 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 147.22 |
| Percent Purity | ≥99% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only