missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1-(1-Naphthyl)ethyl Isocyanate 95.0+%, TCI America™
$197.59 - $659.87
Chemical Identifiers
| CAS | 73671-79-1 |
|---|---|
| Molecular Formula | C13H11NO |
| Molecular Weight (g/mol) | 197.24 |
| MDL Number | MFCD00064181 |
| InChI Key | GONOHGQPZFXJOJ-UHFFFAOYNA-N |
| Synonym | s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate |
| PubChem CID | 7023510 |
| IUPAC Name | 1-(1-isocyanatoethyl)naphthalene |
| SMILES | CC(N=C=O)C1=C2C=CC=CC2=CC=C1 |
Chemical Identifiers
| 73671-79-1 | |
| 197.24 | |
| GONOHGQPZFXJOJ-UHFFFAOYNA-N | |
| 7023510 | |
| CC(N=C=O)C1=C2C=CC=CC2=CC=C1 |
| C13H11NO | |
| MFCD00064181 | |
| s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate | |
| 1-(1-isocyanatoethyl)naphthalene |
Specifications
| 73671-79-1 | |
| C13H11NO | |
| 1 g | |
| s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate | |
| CC(N=C=O)C1=C2C=CC=CC2=CC=C1 | |
| 197.24 | |
| 197.24 | |
| Liquid |
| Yellow | |
| MFCD00064181 | |
| 2206 | |
| GONOHGQPZFXJOJ-UHFFFAOYNA-N | |
| 1-(1-isocyanatoethyl)naphthalene | |
| 7023510 | |
| ≥95.0% (GC) | |
| (S)-(+)-1-(1-Naphthyl)ethyl Isocyanate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator