accessibility menu, dialog, popup

UserName

CAS RN 73671-79-1

IUPAC Name: 1-(1-isocyanatoethyl)naphthalene
Synonyms: 1-(1-isocyanatoethyl)naphthalene
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: GONOHGQPZFXJOJ-UHFFFAOYNA-N
SMILES: CC(N=C=O)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)