missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-(1-Naphthyl)ethyl Isocyanate 95.0+%, TCI America™

Catalog No. I03985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03985G 5 g
I03981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03985G Supplier TCI America Supplier No. I03985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73671-79-1
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00064181
InChI Key GONOHGQPZFXJOJ-UHFFFAOYNA-N
Synonym s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate
PubChem CID 7023510
IUPAC Name 1-(1-isocyanatoethyl)naphthalene
SMILES CC(N=C=O)C1=C2C=CC=CC2=CC=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 2206
Formula Weight 197.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-(1-Naphthyl)ethyl Isocyanate
TSCA No
Recommended Storage Refrigerator