Learn More
(S)-(+)-1-(1-Naphthyl)ethyl isocyanate, 95%
CAS: 73671-79-1 | C13H11NO | 197.24 g/mol
Supplier: Thermo Scientific Chemicals L2024203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (S)-(+)-1-(1-Naphthyl)ethyl isocyanate | |
| 1.118 | |
| 93°C (199°F) | |
| 1.6045 | |
| 1 g | |
| 4675069 | |
| s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate | |
| +47° (c=3.5 in Toluene) | |
| 1-[(1S)-1-isocyanatoethyl]naphthalene | |
| 7023510 | |
| 95% |
| 73671-79-1 | |
| 106°C to 108°C (0.16 mmHg) | |
| C13H11NO | |
| MFCD00064181 | |
| UN2206 | |
| Moisture sensitive; May form a precipitate | |
| GONOHGQPZFXJOJ-UHFFFAOYNA-N | |
| CC(N=C=O)C1=C2C=CC=CC2=CC=C1 | |
| 197.24 | |
| 197.24 |
Chemical Identifiers
| 73671-79-1 | |
| 197.24 | |
| GONOHGQPZFXJOJ-UHFFFAOYNA-N | |
| 7023510 | |
| CC(N=C=O)C1=C2C=CC=CC2=CC=C1 |
| C13H11NO | |
| MFCD00064181 | |
| s-+-1-1-naphthyl ethyl isocyanate, 1-1s-1-isocyanatoethyl naphthalene, s-nei, s-1-1-naphthyl ethyl isocyanate, isocyanic acid s-+-1-1-naphthyl ethyl ester, 1-1-isocyanatoethyl naphthalene #, pubchem8101, s-+-1-naphthyl ethyl isocyanate, s--1-1-naphthyl ethyl isocyanate, s-+-1-1-naphthyl ethylisocyanate | |
| 1-[(1S)-1-isocyanatoethyl]naphthalene |
Safety and Handling
GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only