missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(1-Naphthyl)ethanol 97.0+%, TCI America™

Catalog No. n10635g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
N10635G 5 g
N10631G 1 g
2 options

Catalog No. N10635G

Supplier: TCI America N10635G

Only null left

Chemical Identifiers

CAS 15914-84-8
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00077831
InChI Key CDRQOYRPWJULJN-VIFPVBQESA-N
Synonym s---1-1-naphthyl ethanol, s-1-naphthalen-1-yl-ethanol, s-1-naphthalen-1-yl ethanol, s---1-1-naphtyl ethanol, 1s-1-naphthalen-1-yl ethan-1-ol, s-1-1-naphthyl ethanol, s---alpha-methyl-1-naphthalenemethanol, 1s-1-naphthalen-1-ylethanol, 1s-1-naphthalen-1-yl ethanol, s-+-alpha-methyl-1-naphthalenemethanol
PubChem CID 6976566
IUPAC Name (1S)-1-naphthalen-1-ylethanol
SMILES CC(C1=CC=CC2=CC=CC=C21)O

Specifications

Melting Point 46°C
Boiling Point 165°C
Quantity 5 g
Formula Weight 172.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-1-(1-Naphthyl)ethanol