Learn More
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, 98+%
CAS: 35193-64-7 | C20H13O4P | 348.294 g/mol
Supplier: Thermo Scientific Chemicals L1413706
Description
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
Solubility
Soluble in DMSO and methanol (hot).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
| (S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate | |
| >300°C | |
| MFCD00010045 | |
| 4787775 | |
| Soluble in DMSO and methanol (hot). | |
| +605° (c=1 in Methanol) | |
| 348.294 | |
| 348.3 |
| 35193-64-7 | |
| C20H13O4P | |
| 5 g | |
| 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide | |
| JEHUZVBIUCAMRZ-UHFFFAOYSA-N | |
| C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O | |
| 99589 | |
| ≥98% |
Chemical Identifiers
| 35193-64-7 | |
| 348.294 | |
| JEHUZVBIUCAMRZ-UHFFFAOYSA-N | |
| 99589 |
| C20H13O4P | |
| MFCD00010045 | |
| 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide | |
| C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only