missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate, 98+%
Description
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
Solubility
Soluble in DMSO and methanol (hot).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Used as a chiral Bronsted acid catalyst in the enantoselective Mannich reaction.
Solubility
Soluble in DMSO and methanol (hot).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 1 g |
| Beilstein | 4787775 |
| Solubility Information | Soluble in DMSO and methanol (hot). |
| Optical Rotation | +605° (c=1 in Methanol) |
| Formula Weight | 348.3 |
| Percent Purity | ≥98% |
| Chemical Name or Material | (S)-(+)-1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only