missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,1'-Bi(2-naphthylamine), 97%

CAS: 18531-95-8 | C20H16N2 | 284.362 g/mol

$99.22 - $282.45

Chemical Identifiers

CAS 18531-95-8
Molecular Formula C20H16N2
Molecular Weight (g/mol) 284.362
MDL Number MFCD00145204
InChI Key DDAPSNKEOHDLKB-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
PubChem CID 20571
IUPAC Name 1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH25784MD
View Documents
Thermo Scientific Chemicals
H25784MD
250 mg
Each for $99.22
Only null left
 
AAH2578403
View Documents
Thermo Scientific Chemicals
H2578403
1 g
Each for $282.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18531-95-8
284.362
DDAPSNKEOHDLKB-UHFFFAOYSA-N
20571
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N
C20H16N2
MFCD00145204
1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
1-(2-aminonaphthalen-1-yl)naphthalen-2-amine

Specifications

18531-95-8
C20H16N2
250 mg
1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
−150° (c=1 in Pyridine)
1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
20571
97%
236°C to 242°C
MFCD00145204
2419071
DDAPSNKEOHDLKB-UHFFFAOYSA-N
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N
284.362
284.36
(S)-(-)-1,1'-Bi(2-naphthylamine)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only