accessibility menu, dialog, popup

UserName

CAS RN 18531-95-8

IUPAC Name: [1,1'-binaphthalene]-2,2'-diamine
Synonyms: [1,1'-binaphthalene]-2,2'-diamine
Molecular Weight (g/mol): 284.36
Molecular Formula: C20H16N2
InChi Key: DDAPSNKEOHDLKB-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=CC=CC2=C1C1=C(N)C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)