missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol, TCI America™

Catalog No. T14821G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14821G Supplier TCI America Supplier No. T14821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108998-83-0
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.362
MDL Number MFCD00075492
InChI Key GWVWUZJOQHWMFB-IBGZPJMESA-N
Synonym s---1,1,2-triphenylethane-1,2-diol, s---1,1,2-triphenyl-1,2-ethanediol, s-1,1,2-triphenylethane-1,2-diol, 2s-1,1,2-triphenylethane-1,2-diol, s-1,1,2-triphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2-triphenyl-, 2s, 1,1,2-triphenyl-ethane-1,2-diol, s-1,1,2-triphenyl-ethane-1,2-diol, s-triphenyl-ethane-1,2-diol, s---triphenylethylene glycol
PubChem CID 7000031
IUPAC Name (2S)-1,1,2-triphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)(C3=CC=CC=C3)O)O

Specifications

Melting Point 128°C
Color White
Quantity 1 g
Formula Weight 290.36
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-1,1,2-Triphenyl-1,2-ethanediol