missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,1,2-Triphenyl-1,2-ethanediol, 98%, Thermo Scientific Chemicals

Catalog No. AAH6018906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6018906 5 g
AAH6018903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6018906 Supplier Thermo Scientific Chemicals Supplier No. H6018906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108998-83-0 | Molecular Formula: C20H18O2 | Molecular Weight: 290.362 g/mol

It is used as pharmaceutical and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical and fine chemical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 108998-83-0
Molecular Formula C20H18O2
Molecular Weight (g/mol) 290.362
MDL Number MFCD00075492
InChI Key GWVWUZJOQHWMFB-IBGZPJMESA-N
Synonym s---1,1,2-triphenylethane-1,2-diol, s---1,1,2-triphenyl-1,2-ethanediol, s-1,1,2-triphenylethane-1,2-diol, 2s-1,1,2-triphenylethane-1,2-diol, s-1,1,2-triphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2-triphenyl-, 2s, 1,1,2-triphenyl-ethane-1,2-diol, s-1,1,2-triphenyl-ethane-1,2-diol, s-triphenyl-ethane-1,2-diol, s---triphenylethylene glycol
PubChem CID 7000031
IUPAC Name (2S)-1,1,2-triphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)(C3=CC=CC=C3)O)O

Specifications

Melting Point 125°C to 127°C
Color Brown
Quantity 5 g
Beilstein 3208922
Formula Weight 290.36
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-(-)-1,1,2-Triphenyl-1,2-ethanediol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only