missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RYK Monoclonal Antibody (F35 P7 D7 F5)
GREENER_CHOICE

Catalog No. MA1615
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1615 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1615 Supplier Invitrogen™ Supplier No. MA1615
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

This antibody detects RYK in human, mouse, and rat samples. MA1-615 has been successfully used in Western blot, procedures. By Western blot this antibody detects the RYK receptor-like tyrosine kinase-1 in cells. The MA1-615 immunogen corresponds to the ovalbumin-conjugated synthetic peptide C (462) L T E F H A A L G A Y V(475).

RYK is an atypical member of the family of growth factor receptor protein tyrosine kinases, differing from other members at a number of conserved residues in the activation and nucleotide binding domains. This gene product belongs to a subfamily whose members do not appear to be regulated by phosphorylation in the activation segment. It has been suggested that mediation of biological activity by recruitment of a signaling-competent auxiliary protein may occur through an as yet uncharacterized mechanism. A nine nucleotide insertion in some transcripts results in the SLG variant. It is not established whether this is a product of alternative splicing or a second gene, since evidence for a second gene or pseudogene on chromosome 17 exists.

Specifications

Antigen RYK
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone F35 P7 D7 F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene RYK
Gene Accession No. P34925, Q01887
Gene Alias AW536699; D3S3195; ERK3; ERK-3; growth factor receptor; hydroxyaryl-protein kinase; JTK5; JTK5A; JTK5A protein tyrosine kinase; kinase VIK; Met-related kinase; Mrk; NYK-R; receptor-like tyrosine kinase; RYK; RYK1; tyrosine-protein kinase RYK; Vik
Gene Symbols RYK
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues C(462) L T E F H A A L G A Y V(475) of RYK, conjugated to ovalbumin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140585, 20187, 6259
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less