missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium Standard for AAS, Ready-to-Use, MilliporeSigma™ Supelco™

Catalog No. 111016914
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016914 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016914 Supplier Merck Emd Millipore Supplier No. 84033100ML

Specifications

CAS 7647-01-0
MDL Number MFCD00011207
Synonym Ruthenium(III) chloride solution
Type Analytical Standard
Quantity 100 mL
Grade Analytical Standard