missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium powder, -200 mesh, 99.95% (metals basis)

Catalog No. p-7048921
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1206104 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1206104 Supplier Thermo Scientific Chemicals Supplier No. 01206104
Only null left

CAS: 7440-18-8 | Ru | 101.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Physical Form Powder
Quantity 2 g
UN Number UN3089
Percent Purity 99.95%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Ruthenium powder
Hazard Category H228-H500
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 231-127-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only