missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Ru 75.2% min

CAS: 12036-10-1 | O2Ru | 133.07 g/mol

$97.45 - $97.45

Chemical Identifiers

CAS 12036-10-1
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00011210
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
Synonym ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
PubChem CID 82848
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1180402
View Documents
Thermo Scientific Chemicals
01180402
0.5 g N/A
Only null left
 
Description

Description

The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12036-10-1
133.07
WOCIAKWEIIZHES-UHFFFAOYSA-N
82848
O=[Ru]=O
O2Ru
MFCD00011210
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
dioxoruthenium

Specifications

1200° (sublimation)
Black
0.5 g
O2Ru
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
WOCIAKWEIIZHES-UHFFFAOYSA-N
dioxoruthenium
82848
Odorless
Black powder
Ruthenium(IV) oxide, anhydrous, Ru 75.2% min.
12036-10-1
Powder
99.95% (metals basis)
MFCD00011210
Insoluble in water,and acids. Soluble in fused alkali.
O=[Ru]=O
133.07
133.07
Hygroscopic
6.97
Videos
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 234-840-6

RTECSNumber : VM2654000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only