Learn More
Ruthenium(IV) oxide, anhydrous, 99.9%
Description
The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
Solubility
Insoluble in water, and acids. Soluble in fused alkali.
Notes
Incompatible with oxidizers, acids, aqua regia, organic solvents.
Specifications
Specifications
| Melting Point | 1200°C (sublimation) |
| Quantity | 1 g |
| UN Number | UN1479 |
| Solubility Information | Insoluble in water,and acids. Soluble in fused alkali. |
| Formula Weight | 133.07 |
| Percent Purity | 99.9% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 6.97 |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H319 |
| Hazard Statement | GHS H Statement H271 May cause fire or explosion strong oxidizer. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S. |
| EINECSNumber | 234-840-6 |
| RTECSNumber | VM2654000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only