missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosylchloride hydrate

Catalog No. AA4308903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4308903 1 g
AA4308906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4308903 Supplier Thermo Scientific Chemicals Supplier No. 04308903

CAS: 32535-76-5 | Cl3NORu | 237.42 g/mol

Used as catalyst. As a precursor to other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32535-76-5
Molecular Formula Cl3NORu
Molecular Weight (g/mol) 237.42
MDL Number MFCD00049765
InChI Key XTHALOZYNGMAKY-UHFFFAOYSA-K
IUPAC Name trichloro(nitroso)rutheniumtris(ylium)
SMILES Cl[Ru+3](Cl)(Cl)N=O

Specifications

Melting Point 180°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 237.44 (Anhydrous)
Chemical Name or Material Ruthenium (III) nitrosylchloride hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only