accessibility menu, dialog, popup

UserName

ClRuClNClO

CAS RN 32535-76-5

IUPAC Name: trichloro(nitroso)rutheniumtris(ylium)
Synonyms: trichloro(nitroso)rutheniumtris(ylium)
Molecular Weight (g/mol): 237.42
Molecular Formula: Cl3NORu
InChi Key: XTHALOZYNGMAKY-UHFFFAOYSA-K
SMILES: Cl[Ru+3](Cl)(Cl)N=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results