missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) iodide, anhydrous, Ru 20.5% min

Catalog No. AA1180503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1180503 1 g
AA1180506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1180503 Supplier Thermo Scientific Chemicals Supplier No. 01180503
Only null left

CAS: 13896-65-6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13896-65-6
MDL Number MFCD00016316
Synonym ruthenium iii iodide, ruthenium iii ; iodide, ruthenium 3+ triiodide, ruthenium iii iodide anhydrous, ruthenium iii iodide, anhydrous 1g, ruthenium iii iodide, anhydrous, ru min

Specifications

Melting Point 590°C (decomposition)
Physical Form Powder
Quantity 1 g
Formula Weight 481.78
Concentration or Composition (by Analyte or Components) Ru 20.5% min.
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) iodide, Anhydrous
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-664-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only