missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) Chloride, TCI America™

Catalog No. R00741G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00741G Supplier TCI America Supplier No. R00741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ruthenium-based catalyst used for the synthesis of unsaturated N-glycosides via ferrier azaglycosylation

  • Anhydrous ruthenium content of min 40.0 % to be stored under inert gas

Chemical Identifiers

CAS 10049-08-8
Molecular Formula Cl3Ru
MDL Number MFCD00011208
Synonym Ruthenium Trichloride Hydrate, Trichlororuthenium Hydrate

Specifications

Color Black
Physical Form Crystalline Powder
Quantity 1 g
Linear Formula RuCl3⋅xH2O
UN Number 3260
Formula Weight 207.42
Loss on Drying 1.0 to 5.0%
Packaging Glass Bottle With Plastic Inert
Sensitivity Hygroscopic
Appearance Black Powder to Crystal
Chemical Name or Material Ruthenium(III) Chloride
Show More Show Less
EINECSNumber (1)-0650
RTECSNumber VM2650000
TSCA Yes