missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, AAS standard solution, Specpure™ Ru 1000μg/mL

$108.64 - $108.64

Chemical Identifiers

MDL Number MFCD00011207
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA89117AE
View Documents
Thermo Scientific Chemicals
089117AE
100 mL
Each for $108.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00011207

Specifications

Liquid
MFCD00011207
100 mL
Ruthenium, AAS standard solution
Ru 1000 μg/mL; RuCl3 in 20% HCl
UN1789
Specpure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID

EINECSNumber : 231-595-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only