missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rutaecarpine 98.0+%, TCI America™
$369.08 - $369.08
Chemical Identifiers
| CAS | 84-26-4 |
|---|---|
| Molecular Formula | C18H13N3O |
| Molecular Weight (g/mol) | 287.32 |
| MDL Number | MFCD00210551 |
| InChI Key | ACVGWSKVRYFWRP-UHFFFAOYSA-N |
| Synonym | 8,13-Dihydroindolo[2′C,3′C:3,4]pyrido[2,1-b]quinazolin-5(7H)-one |
| PubChem CID | 65752 |
| ChEBI | CHEBI:8922 |
| IUPAC Name | 3,13,21-triazapentacyclo[11.8.0.0²,¹â°.0â´,â¹.0¹âµ,²â°]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one |
| SMILES | O=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0102200MG
|
TCI America
R0102200MG |
200 mg |
N/A
|
|
|||||
Chemical Identifiers
| 84-26-4 | |
| 287.32 | |
| ACVGWSKVRYFWRP-UHFFFAOYSA-N | |
| 65752 | |
| 3,13,21-triazapentacyclo[11.8.0.0²,¹â°.0â´,â¹.0¹âµ,²â°]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one |
| C18H13N3O | |
| MFCD00210551 | |
| 8,13-Dihydroindolo[2′C,3′C:3,4]pyrido[2,1-b]quinazolin-5(7H)-one | |
| CHEBI:8922 | |
| O=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12 |
Specifications
| 84-26-4 | |
| Yellow | |
| MFCD00210551 | |
| 8,13-Dihydroindolo[2′C,3′C:3,4]pyrido[2,1-b]quinazolin-5(7H)-one | |
| O=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12 | |
| 287.32 | |
| CHEBI:8922 | |
| ≥98.0% (HPLC,N) | |
| Rutaecarpine |
| 262°C | |
| C18H13N3O | |
| 200 mg | |
| ACVGWSKVRYFWRP-UHFFFAOYSA-N | |
| 3,13,21-triazapentacyclo[11.8.0.0²,¹â°.0â´,â¹.0¹âµ,²â°]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one | |
| 65752 | |
| 287.32 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : NM3454000
TSCA : No
Recommended Storage : Refrigerator