missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ RUNX1 Plasmid

Catalog No. NBP22528110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22528110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22528110 Supplier R&D Systems™ Supplier No. NBP225281100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Mammalian expression vector construct, pCMV/hRUNX1, containing coding sequence of human Runt-related transcription factor 1

The RUNX1 Plasmid is a mammalian expression vector construct, pCMV/hRUNX1, containing coding sequence of human Runt-related transcription factor 1 (GenBank Accession: BC136381). Gene ID: 861

Specifications

Concentration 0.5mg/mL
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
For Use With (Application) Expression of hRUNX1 protein in mammalian cells. Induction of FOXP3 promoter activity. Total content of 10μg is enough for one 96-well cell transfection (0.1μg plasmid per well). Recommended amount for other plate formats: 6-well plate (2μg/well), 12-well plate (0.8μg/well) and 24-well plate (0.4μg/well).
Product Line Novus
Product Type RUNX1 Plasmid - Target
Quantity 100 μg
Species Human (Hu)