missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ RUNX1 Plasmid

Catalog No. NBP225281
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225281 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP225281 Supplier R&D Systems™ Supplier No. NBP225281

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. For expression of hRUNX1 protein in mammalian cells, induction of FOXP3 promoter activity

The RUNX1 Plasmid is a mammalian expression vector construct, pCMV/hRUNX1, containing coding sequence of human Runt-related transcription factor 1.

  • Species: Human
  • Gene: RUNX1
  • Immunogen: RUNX1 Plasmid
  • Total contents of 10μg is enough for one 96-well cell transfection

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Line Novus
Product Type RUNX1 Plasmid
Quantity 10 μg