missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1/CBFA2 Antibody, R&D Systems™
Description
RUNX1/CBFA2 Polyclonal specifically detects RUNX1/CBFA2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | RUNX1/CBFA2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2-5 ug/ml |
| Formulation | PBS containing 0.2% gelatin with 0.05% Sodium Azide |
| Gene Accession No. | Q01196 |
| Gene Alias | Acute myeloid leukemia 1 protein, AML1acute myeloid leukemia 1, AML1-EVI-1, CBFA2acute myeloid leukemia 1 protein (oncogene AML-1), core-binding factor, alphasubunit10AMLCR1, Core-binding factor subunit alpha-2, EVI-1, PEBP2A2, runt domain, alpha subunit 2, runt-related transcription factor 1, SL3/AKV core-binding factor alpha B subunit |
| Gene Symbols | RUNX1 |
| Show More |
For Research Use Only