missing translation for 'onlineSavingsMsg'
Learn More

RuCl[(S,S)-Fsdpen](p-cymene) 90.0+%, TCI America™

Catalog No. p-7135705
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0122200MG 200 mg
R01221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0122200MG Supplier TCI America Supplier No. R0122200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1026995-72-1
Molecular Formula C30H29ClF5N2O2RuS+
Molecular Weight (g/mol) 713.146
MDL Number MFCD12545954
InChI Key WILRHPOBOUTKKV-NWMPYMMKSA-M
Synonym [(S,S)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulfonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(pentafluorobenzenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 132274302
IUPAC Name N-[(1S,2S)-2-amino-1,2-diphenylethyl]-2,3,4,5,6-pentafluorobenzenesulfonamide;chlororuthenium(1+);1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)NS(=O)(=O)C3=C(C(=C(C(=C3F)F)F)F)F)N.Cl[Ru+]

Specifications

Melting Point 244°C
Quantity 200 mg
Formula Weight 712.14
Percent Purity ≥90% (NMR)
Physical Form Crystalline Powder
Chemical Name or Material RuCl[(S,S)-Fsdpen](p-cymene)