accessibility menu, dialog, popup

UserName

CAS RN 1026995-72-1

IUPAC Name: 1-methyl-4-(propan-2-yl)benzene; N-(2-amino-1,2-diphenylethyl)-2,3,4,5,6-pentafluorobenzene-1-sulfonamide; chlororutheniumylium
Synonyms: N-(2-amino-1,2-diphenylethyl)-2,3,4,5,6-pentafluorobenzenesulfonamide; chlororutheniumylium; cymene
Molecular Weight (g/mol): 713.15
Molecular Formula: C30H29ClF5N2O2RuS
InChi Key: WILRHPOBOUTKKV-UHFFFAOYNA-M
SMILES: Cl[Ru+].CC(C)C1=CC=C(C)C=C1.NC(C(NS(=O)(=O)C1=C(F)C(F)=C(F)C(F)=C1F)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results