missing translation for 'onlineSavingsMsg'
Learn More

RuCl[(R,R)-Tsdpen](mesitylene) 90.0+%, TCI America™

Catalog No. p-7135710
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0127200MG 200 mg
R01271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0127200MG Supplier TCI America Supplier No. R0127200MG
Only null left

Chemical Identifiers

CAS 174813-82-2
Molecular Formula C30H34ClN2O2RuS
Molecular Weight (g/mol) 623.194
MDL Number MFCD12545951
InChI Key BABSEZBJIPZYQB-AGEKDOICSA-M
Synonym [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(mesitylene)ruthenium(II), Chloro(mesitylene)[(R,R)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 131675324
IUPAC Name N-[(1R,2R)-2-amino-1,2-diphenylethyl]-4-methylbenzenesulfonamide;ruthenium(1+);1,3,5-trimethylbenzene;chloride
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(C2=CC=CC=C2)C(C3=CC=CC=C3)N.CC1=CC(=CC(=C1)C)C.[Cl-].[Ru+]

Specifications

Quantity 200 mg
Formula Weight 622.19
Percent Purity ≥90% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material RuCl[(R,R)-Tsdpen](mesitylene)