accessibility menu, dialog, popup

UserName

CAS RN 174813-82-2

CH3SOONNHCH3H3CCH3ClRu

CAS RN 174813-82-2

IUPAC Name: (2-azanidyl-1,2-diphenylethyl)(4-methylbenzenesulfonyl)azanide; 1,3,5-trimethylbenzene; chlororutheniumbis(ylium)
Synonyms: (2-azanidyl-1,2-diphenylethyl)(4-methylbenzenesulfonyl)azanide; chlororutheniumbis(ylium); mesitylene
Molecular Weight (g/mol): 621.18
Molecular Formula: C30H32ClN2O2RuS
InChi Key: GSFQTYRZIWRZCD-UHFFFAOYNA-M
SMILES: Cl[Ru++].CC1=CC(C)=CC(C)=C1.CC1=CC=C(C=C1)S(=O)(=O)[N-]C(C([NH-])C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results