missing translation for 'onlineSavingsMsg'
Learn More

[RuCl(p-cymene)((S)-binap)]Cl 90.0+%, TCI America™

Catalog No. p-7135726
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0147200MG 200 mg
R01471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0147200MG Supplier TCI America Supplier No. R0147200MG
Only null left

Chemical Identifiers

CAS 130004-33-0
Molecular Formula C54H46Cl2P2Ru
Molecular Weight (g/mol) 928.88
MDL Number MFCD00134456
InChI Key WNHLGYRPKARUHY-UHFFFAOYSA-L
Synonym chloro s---2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride, r-+-2,2-bis diphenylphosphino-1,1-binaphthalenechloro p-cymene ruthenium chloride, chloro r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl p-cymene ruthenium ii chloride, rucl p-cymene s-binap cl, r-2,2'-bis-diphenylphosphino-1,1'-binaphthyl-chloro-p-cymene-rutheniumchloride, s-2,2'-bis-diphenylphosphino-1,1'-binaphthyl-chloro-p-cymene-rutheniumchloride
PubChem CID 11147374
IUPAC Name dichlororuthenium;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl[Ru]Cl

Specifications

Quantity 200 mg
Formula Weight 928.88
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [RuCl(p-cymene)((S)-binap)]Cl