missing translation for 'onlineSavingsMsg'
Learn More

Rubidium sulfate, 99% (metals basis)

CAS: 7488-54-2 | H2O4RbS | 183.54 g/mol

$2773.30 - $2773.30

Chemical Identifiers

CAS 7488-54-2
Molecular Formula H2O4RbS
Molecular Weight (g/mol) 183.54
MDL Number MFCD00011190
InChI Key XNNUAZGCOYTNQE-UHFFFAOYSA-N
PubChem CID 197088
IUPAC Name sulfuric acid rubidium
SMILES [Rb].OS(O)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1349336
View Documents
Thermo Scientific Chemicals
01349336
500 g
Each for $2,773.30
Only null left
 
Description

Description

Rubidium sulfate is used in hematology stain. It is also used as the raw materials for making kinds of Rubidium salts. It is fused on a ceramic bead and used as thermoionic detector. It is also used as a cathartic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7488-54-2
183.54
XNNUAZGCOYTNQE-UHFFFAOYSA-N
sulfuric acid rubidium
H2O4RbS
MFCD00011190
197088
[Rb].OS(O)(=O)=O

Specifications

1050°C
Crystalline
H2O4RbS
rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate
XNNUAZGCOYTNQE-UHFFFAOYSA-N
sulfuric acid rubidium
197088
99%
1.513
Rubidium sulfate
7488-54-2
500 g
MFCD00011190
Soluble in water (424mg/ml at 10°C).
[Rb].OS(O)(=O)=O
183.54
267
Odorless
3.613 g/mL
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 231-301-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only