accessibility menu, dialog, popup

UserName

CAS RN 7488-54-2

IUPAC Name: sulfuric acid rubidium
Synonyms: sulfuric acid rubidium
Molecular Weight (g/mol): 183.54
Molecular Formula: H2O4RbS
InChi Key: XNNUAZGCOYTNQE-UHFFFAOYSA-N
SMILES: [Rb].OS(O)(=O)=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)