missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rubidium chloride, 99.99%, (trace metal basis), Thermo Scientific™
Chemical Identifiers
| CAS | 7791-11-9 |
|---|---|
| Molecular Formula | ClRb |
| Molecular Weight (g/mol) | 120.92 |
| InChI Key | FGDZQCVHDSGLHJ-UHFFFAOYSA-M |
| Synonym | rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride |
| PubChem CID | 62683 |
| ChEBI | CHEBI:78672 |
| IUPAC Name | rubidium(1+);chloride |
| SMILES | [Cl-].[Rb+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC193920100
|
Thermo Scientific Chemicals
193920100 |
10g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For ultra trace analysisChemical Identifiers
| 7791-11-9 | |
| 120.92 | |
| rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride | |
| CHEBI:78672 | |
| [Cl-].[Rb+] |
| ClRb | |
| FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
| 62683 | |
| rubidium(1+);chloride |
Specifications
| 7791-11-9 | |
| 28.9 to 30.1% (Cl) (Argentometry) | |
| RbCl | |
| rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride | |
| FGDZQCVHDSGLHJ-UHFFFAOYSA-M | |
| rubidium(1+);chloride | |
| 62683 | |
| 120.92 | |
| Trace Metal Basis | |
| Rubidium chloride |
| 10g | |
| ClRb | |
| 15,8417 | |
| Solubility in water: 910g/L (20 c). Other solubilities: 1g/0.7mL boiling water,practically insoluble in alcohols | |
| [Cl-].[Rb+] | |
| 120.92 | |
| CHEBI:78672 | |
| 99.99% | |
| Glass bottle |
Safety and Handling
EINECSNumber : 232-240-9
RUO â Research Use Only