missing translation for 'onlineSavingsMsg'
Learn More

Rubidium Bromide, Approx. 99% -, Spectrum™ Chemical

$265.78 - $1953.09

Chemical Identifiers

CAS 7789-39-1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613049
View Documents
Spectrum Chemical Mfg Cor
R13585GM
5 g Amber Glass Bottle
Each for $265.78
Only null left
 
18613047
View Documents
Spectrum Chemical Mfg Cor
R1358100GM
100 g Amber Glass Bottle
Each for $1,953.09
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7789-39-1

Specifications

7789-39-1
5 g
Reagent
1
∼99%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.