missing translation for 'onlineSavingsMsg'
Learn More

Rubidium Bromide, Approx. 99% -, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor R13585GM

 View more versions of this product

Catalog No. 18613049


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7789-39-1
5 g
Reagent
1
∼99%
Amber Glass Bottle

Chemical Identifiers

7789-39-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.