missing translation for 'onlineSavingsMsg'
Learn More

Rubiadin, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012659
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012659 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012659 Supplier Merck Emd Millipore Supplier No. 7455310MG
Food and Beverages

Specifications

CAS 117-02-2
Physical Form Neat
Linear Formula C15H10O4
MDL Number MFCD02752095
Synonym 1,3-Dihydroxy-2-methyl-9,10-anthracenedione; 1,3-Dihydroxy-2-methylanthraquinone
Type Analytical Standard
Quantity 10 mg
Formula Weight 254.24
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard