Learn More
Eisco™ Rubber Cones
Designed for use with Buchner Funnels in filtration assemblies, these cones are made of flexible, quality neoprene rubber. Rubber Cones have superior chemical stability, and can withstand a wide temperature range.
Supplier:  Eisco™ CH0421C
 View more versions of this product
Catalog No. S27054
Only null left
Description
Description
  • Tapered filter adapter cone
  • Made of flexible, and quality neoprene rubber
  • Good chemical stability
  • Able to withstand a wide temperature range
  • Designed for use with Buchner funnels in filtration assemblies
  • Can be nested within different size cones for convenient storage
  • Autoclavable
Specifications
Specifications
Non-sterile
Rubber Stopper
3 mm
36 mm
3.81 cm
25 mm
Blue
3
23.5 mm
Neoprene
Not Autoclavable
Conical
Product Suggestions
Product Suggestions