missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ru(OAc)2[(R)-tolbinap], TCI America™
$74.56 - $246.05
Chemical Identifiers
| CAS | 116128-29-1 |
|---|---|
| Molecular Formula | C52H48O4P2Ru |
| Molecular Weight (g/mol) | 899.97 |
| MDL Number | MFCD09753021,MFCD09753021 |
| InChI Key | OXESSJMVRGFBNX-UHFFFAOYSA-N |
| Synonym | Diacetato[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]ruthenium(II) |
| PubChem CID | 85364800 |
| IUPAC Name | bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium |
| SMILES | [Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0168200MG
|
TCI America
R0168200MG |
200 mg |
Each for $74.56
|
|
|||||
|
R01681G
|
TCI America
R01681G |
1 g |
Each for $246.05
|
|
|||||
Chemical Identifiers
| 116128-29-1 | |
| 899.97 | |
| OXESSJMVRGFBNX-UHFFFAOYSA-N | |
| 85364800 | |
| [Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1 |
| C52H48O4P2Ru | |
| MFCD09753021,MFCD09753021 | |
| Diacetato[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]ruthenium(II) | |
| bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium |
Specifications
| 116128-29-1 | |
| C52H48O4P2Ru | |
| 200 mg | |
| OXESSJMVRGFBNX-UHFFFAOYSA-N | |
| bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium | |
| 85364800 | |
| Crystalline Powder |
| Black-Yellow | |
| MFCD09753021,MFCD09753021 | |
| Diacetato[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]ruthenium(II) | |
| [Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1 | |
| 899.97 | |
| 897.95 | |
| Ru(OAc)2[(R)-tolbinap] |
Safety and Handling
TSCA : No