missing translation for 'onlineSavingsMsg'
Learn More

Ru(OAc)2[(R)-tolbinap], TCI America™

Catalog No. R0168200MG
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0168200MG 200 mg
R01681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. R0168200MG missing translation for 'mfr' TCI America missing translation for 'supplierNo' R0168200MG

Chemical Identifiers

CAS 116128-29-1
Molecular Formula C52H48O4P2Ru
Molecular Weight (g/mol) 899.97
MDL Number MFCD09753021,MFCD09753021
InChI Key OXESSJMVRGFBNX-UHFFFAOYSA-N
Synonym Diacetato[(R)-(+)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl]ruthenium(II)
PubChem CID 85364800
IUPAC Name bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium
SMILES [Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Color Black-Yellow
Quantity 200 mg
Formula Weight 897.95
Physical Form Crystalline Powder
Chemical Name or Material Ru(OAc)2[(R)-tolbinap]