missing translation for 'onlineSavingsMsg'
Learn More

Ru-MACHO(regR) (contains 5% Toluene at maximum), TCI America™

Catalog No. R0136200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0136200MG 200 mg
R01361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0136200MG Supplier TCI America Supplier No. R0136200MG
Only null left

Chemical Identifiers

CAS 1295649-40-9
Molecular Formula C29H30ClNOP2Ru
Molecular Weight (g/mol) 607.033
MDL Number MFCD22377801
InChI Key YCZWZQUMPDDGQF-UHFFFAOYSA-M
Synonym Carbonylchlorohydrido[bis(2-diphenylphosphinoethyl)amino]ruthenium(II)
PubChem CID 132774591
IUPAC Name carbon monoxide;chloro(hydrido)ruthenium;2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine
SMILES [C-]#[O+].C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl[RuH]

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 607.03
Physical Form Crystalline Powder
Chemical Name or Material Ru-MACHO(regR) (contains 5% Toluene at maximum)