missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Ru-MACHO(regR)-BH | |
| White-Yellow | |
| MFCD20922901 | |
| Carbonylhydrido(tetrahydroborato)[bis(2-diphenylphosphinoethyl)amino]ruthenium(II) | |
| [BH4-].[RuH+].[C-]#[O+].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 586.42 | |
| 586.42 |
| 1295649-41-0 | |
| C29H34BNOP2Ru | |
| 1 g | |
| ZVKXESLAUDGPND-UHFFFAOYSA-N | |
| λ²-ruthenium(1+) bis[2-(diphenylphosphanyl)ethyl]amine methanidylidyneoxidanium boranuide | |
| 132774516 | |
| Crystalline Powder |
Chemical Identifiers
| 1295649-41-0 | |
| 586.42 | |
| ZVKXESLAUDGPND-UHFFFAOYSA-N | |
| 132774516 | |
| [BH4-].[RuH+].[C-]#[O+].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1 |
| C29H34BNOP2Ru | |
| MFCD20922901 | |
| Carbonylhydrido(tetrahydroborato)[bis(2-diphenylphosphinoethyl)amino]ruthenium(II) | |
| λ²-ruthenium(1+) bis[2-(diphenylphosphanyl)ethyl]amine methanidylidyneoxidanium boranuide |
Safety and Handling
TSCA : No