missing translation for 'onlineSavingsMsg'
Learn More

Ru-MACHO(regR)-BH, TCI America™

Catalog No. p-7135720
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0137200MG 200 mg
R01371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0137200MG Supplier TCI America Supplier No. R0137200MG
Only null left

Chemical Identifiers

CAS 1295649-41-0
Molecular Formula C29H34BNOP2Ru
Molecular Weight (g/mol) 586.42
MDL Number MFCD20922901
InChI Key ZVKXESLAUDGPND-UHFFFAOYSA-N
Synonym Carbonylhydrido(tetrahydroborato)[bis(2-diphenylphosphinoethyl)amino]ruthenium(II)
PubChem CID 132774516
IUPAC Name λ²-ruthenium(1+) bis[2-(diphenylphosphanyl)ethyl]amine methanidylidyneoxidanium boranuide
SMILES [BH4-].[RuH+].[C-]#[O+].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 586.42
Physical Form Crystalline Powder
Chemical Name or Material Ru-MACHO(regR)-BH