Learn More
Brady™ RTK Pictogram Labels, Poison
Effectively convey personal protection instructions and special precautionary information. Brady™ RTK Pictogram Labels are made from premium grade B-946 vinyl and are sized to fit standard blank write-on right-to-know labels.
Supplier:  Brady™ 58575
 View more versions of this product
Catalog No. 19107228
Only null left
Description
Description
  • Made from premium grade B-946 vinyl
  • Sized to fit standard Blank Write-On Right-To-Know Labels
  • Correspond to ANSI proposed Z.535 Standard
  • Card-mounted for easy removal
Specifications
Chemical Identifiers
Acrylic, Permanent, Cold-temperature
Pressure-sensitive Vinyl (B-946)
0.004 in.
White, Blue
Poison
Specifications
Acrylic, Permanent, Cold-temperature
Pressure-sensitive Vinyl (B-946)
Poison
0.004 in.
White, Blue
RTK (Right-To-Know) Pictogram Label
0.75 x 0.75 in.
Product Suggestions
Product Suggestions